2-Fluor-4-methylbenzylamin 97%

Lieferant: Thermo Scientific

Gefahr

H26267.MDEA 55.4 EUR
H26267.MD H26267.03
2-Fluor-4-methylbenzylamin 97%
2-Fluor-4-methylbenzylamin
Formel: C₈H₁₀FN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06213635
CAS-Nummer: 771573-01-4
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR