3-(Difluormethoxy)benzylamin 98%

Lieferant: Thermo Scientific
Gefahr

H50322.03EA 126 EUR
H50322.03 H50322.06
3-(Difluormethoxy)benzylamin 98%
3-(Difluormethoxy)benzylamin
Formel: C₈H₉F₂NO
MDL Number: MFCD00236230
CAS-Nummer: 244022-71-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR