N,N-Dimethylformamid-[D1] (98% D), Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ200

SIAL392901-1GEA 172 EUR
SIAL392901-1G
N,N-Dimethylformamid-[D1] (98% D), Sigma-Aldrich®
N,N-Dimethylformamid-[D1]
Formel: DCOON(CH₃)₂
CAS-Nummer: 2914-27-4
UN-Nummer: 2265
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR