2,4-Dichlor-3-ethyl-6-nitrophenol
Lieferant: Molekula
MOLE47876479-5GEA
0
EUR
MOLE47876479-5G
2,4-Dichlor-3-ethyl-6-nitrophenol
2,4-Dichlor-3-ethyl-6-nitrophenol
Formel:
C₈H₇Cl₂NO₃ Molekulargewicht: 236,05 g/mol |
MDL Number:
MFCD00270764 CAS-Nummer: 99817-36-4 EINECS: 420-740-1 UN-Nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...