Trichromdicarbid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL402680-50GEA
93.7
EUR
SIAL402680-50G
Trichromdicarbid, Sigma-Aldrich®
Trichromium dicarbide
Formel:
C₂Cr₃ Molekulargewicht: 180,01 g/mol |
MDL Number:
MFCD00016041 CAS-Nummer: 12012-35-0 EINECS: 234-576-1 UN-Nummer: 3178 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...