3-Methyl-5-(trifluormethyl)benzylbromid ≥97%
|
Gefahr
|
H33520.03EA
137
EUR
H33520.03
H33520.MD
3-Methyl-5-(trifluormethyl)benzylbromid ≥97%
3-Methyl-5-(trifluormethyl)benzylbromid
Formel:
C₉H₈BrF₃ Molekulargewicht: 253,06 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01631610 CAS-Nummer: 116070-36-1 UN-Nummer: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...