Triphenylantimondichlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ100

SIAL135097-5GEA 199 EUR
SIAL135097-5G
Triphenylantimondichlorid, Sigma-Aldrich®
Triphenylantimondichlorid
Formel: C₁₈H₁₅Cl₂Sb
Molekulargewicht: 423,98 g/mol
MDL Number: MFCD00013598
CAS-Nummer: 594-31-0
EINECS: 209-834-1
UN-Nummer: 2923
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR