3'-Fluor-5'-(trifluormethoxy)acetophenon ≥97%

Lieferant: Alfa Aesar
Achtung

H34207.03EA 137 EUR
H34207.03 H34207.MD
3'-Fluor-5'-(trifluormethoxy)acetophenon ≥97%
3'-Fluor-5'-(trifluormethoxy)acetophenon
Formel: C₉H₆F₄O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18783230
CAS-Nummer: 1352999-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR