6-Hydroxy-m-anisidin, Sigma-Aldrich®

Lieferant: Merck

SIAL722367-1GEA 62.8 EUR
SIAL722367-1G
6-Hydroxy-m-anisidin, Sigma-Aldrich®
6-Hydroxy-m-anisidin
Formel: C₇H₉NO₂
Molekulargewicht: 139,16
MDL Number: MFCD06616911
CAS-Nummer: 20734-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR