3',5'-Dibrom-4'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL542245-5GEA 54.1 EUR
SIAL542245-5G
3',5'-Dibrom-4'-hydroxyacetophenon, Sigma-Aldrich®
3',5'-Dibrom-4'-hydroxyacetophenon
Formel: C₈H₆Br₂O₂
EINECS: 220-750-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR