2-(Difluoromethoxy)phenylacetonitrile, 97%

H50641.03EA 103 EUR
ALFAH50641.03 ALFAH50641.06
2-(Difluoromethoxy)phenylacetonitrile, 97%
Purity: 97%
CAS No.: 41429-22-5
Molecular Formula: C9H7F2NO
Formula Weight: 183.16
UN No.: 3276
MDL No.: MFCD00236240
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR