2-Methyl-5-(trifluoromethyl)benzamide, 97%

H31516.06EA 130 EUR
ALFAH31516.06 ALFAH31516.03
2-Methyl-5-(trifluoromethyl)benzamide, 97%
Purity: 97%
CAS No.: 261951-97-7
Molecular Formula: C9H8F3NO
Formula Weight: 203.17
MDL No.: MFCD01631496
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR