2'-Methyl-3'-(trifluormethyl)acetophenon ≥97%
|
Achtung
|
H32888.03EA
83.7
EUR
H32888.03
H32888.06
2'-Methyl-3'-(trifluormethyl)acetophenon ≥97%
2'-Methyl-3'-(trifluormethyl)acetophenon
Formel:
C₁₀H₉F₃O Molekulargewicht: 202,17 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832283 CAS-Nummer: 1017778-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...