2-Fluoro-N-isopropylbenzylamine, 97%

H56639.03EA 146 EUR
ALFAH56639.03 ALFAH56639.MD
2-Fluoro-N-isopropylbenzylamine, 97%
Purity: 97%
CAS No.: 921074-63-7
Molecular Formula: C10H14FN
Formula Weight: 167.22
UN No.: 2735
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR