3-Fluoro-N-isopropylbenzylamine, 97%

H56383.03EA 172 EUR
ALFAH56383.03 ALFAH56383.MD
3-Fluoro-N-isopropylbenzylamine, 97%
Purity: 97%
CAS No.: 90389-87-0
Molecular Formula: C10H14FN
Formula Weight: 167.22
UN No.: 2735
MDL No.: MFCD08060644
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR