3-Fluoro-N-n-hexylbenzylamine, 97%

H57777.MDEA 33 EUR
ALFAH57777.MD ALFAH57777.03
3-Fluoro-N-n-hexylbenzylamine, 97%
Purity: 97%
CAS No.: 1042511-87-4
Molecular Formula: C13H20FN
Formula Weight: 209.31
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR