4-Fluoro-N-n-pentylbenzylamine, 97%

H57534.03EA 286 EUR
ALFAH57534.03 ALFAH57534.MD
4-Fluoro-N-n-pentylbenzylamine, 97%
Purity: 97%
CAS No.: 847744-11-0
Molecular Formula: C12H18FN
Formula Weight: 195.28
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR