2,2,2,3',4'-Pentafluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL514624-1GEA 255 EUR
SIAL514624-1G
2,2,2,3',4'-Pentafluoracetophenon, Sigma-Aldrich®
2,2,2,3',4'-Pentafluoracetophenon
Formel: C₈H₃F₅O
MDL Number: MFCD01319992
CAS-Nummer: 302912-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR