7-Fluorchinaldin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL566047-5GEA 56.3 EUR
SIAL566047-5G
7-Fluorchinaldin, Sigma-Aldrich®
7-Fluorchinaldin
Formel: C₁₀H₈FN
MDL Number: MFCD02683029
CAS-Nummer: 1128-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR