Isochinolin-5-sulfonsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL246123-5GEA 48.5 EUR
SIAL246123-5G SIAL246123-25G
Isochinolin-5-sulfonsäure, Sigma-Aldrich®
Isochinolin-5-sulfonsäure
Formel: C₉H₇NO₃S
Molekulargewicht: 209,22 g/mol
MDL Number: MFCD00134089
CAS-Nummer: 27655-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR