Lanthan(III)carbonat Hydrat, Sigma-Aldrich®
Lieferant: Merck
SIAL325767-100GEA
77.4
EUR
SIAL325767-100G
Lanthan(III)carbonat Hydrat, Sigma-Aldrich®
Lanthan(III)carbonat Hydrat
Formel:
La₂(CO₃)₃.xH₂O Molekulargewicht: 457,85 g/mol (anhy) |
MDL Number:
MFCD00011069 CAS-Nummer: 54451-24-0 EINECS: 209-599-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...