3-Cyanchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL383481-5GEA 76.1 EUR
SIAL383481-5G
3-Cyanchromon, Sigma-Aldrich®
3-Cyanchromon
Formel: C₁₀H₅NO₂
Molekulargewicht: 171,16 g/mol
MDL Number: MFCD00052604
CAS-Nummer: 50743-17-4
UN-Nummer: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR