2-(3-Methyl-1-piperidinyl)phenol

Lieferant: Thermo Scientific

S08914.250 S08914.1
CHMPS08914.250EA 0 EUR
CHMPS08914.250 MAYBS08914.1
2-(3-Methyl-1-piperidinyl)phenol
2-(3-Methyl-1-piperidinyl)phenol
Formel: C₁₂H₁₇NO
CAS-Nummer: 78864-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR