(R)-(-)-2-Pentanol ≥97%
|
Gefahr
|
J63298.03EA
221
EUR
J63298.03
(R)-(-)-2-Pentanol ≥97%
(R)-(-)-2-Pentanol
Formel:
C₅H₁₂O Molekulargewicht: 88,15 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00065953 CAS-Nummer: 31087-44-2 UN-Nummer: 1105 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...