BES sodium salt

J64052.14EA 46.5 EUR
ALFAJ64052.14 ALFAJ64052.22
BES sodium salt
CAS No.: 66992-27-6
Molecular Formula: C6H14NNaO5S
Formula Weight: 235.23
Physical Form: Powder
Appearance: White
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR