3-(Trifluoromethyl)benzamidoxime, 97%

H58744.04EA 199 EUR
ALFAH58744.04 ALFAH58744.09
3-(Trifluoromethyl)benzamidoxime, 97%
Purity: 97%
CAS No.: 40067-80-9
Molecular Formula: C8H7N2OF3
Formula Weight: 204.15
Physical Form: Solid
Appearance: Pale brown
Melting Point: 82-88°
MDL No.: MFCD02647312
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR