4-(Trifluoromethyl)benzamidoxime, 97%

H58755.04EA 226 EUR
ALFAH58755.04 ALFAH58755.09
4-(Trifluoromethyl)benzamidoxime, 97%
Purity: 97%
CAS No.: 22179-86-8
Molecular Formula: C8H7F3N2O
Formula Weight: 204.15
Physical Form: Solid
Appearance: Pale brown
Melting Point: 125-130°
MDL No.: MFCD00053042
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR