6-Chlorpicolinsäure
Lieferant: EHRENSTORFER GMBH
EHERC11495000EA
589
EUR
EHERC11495000
6-Chlorpicolinsäure
6-Chlorpicolinsäure
Formel:
C₆H₄ClNO₂ Molekulargewicht: 157,56 g/mol |
MDL Number:
MFCD00155390 CAS-Nummer: 4684-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...