MSIA pipette tips
Lieferant: Thermo Scientific
732-1646EA 347 EUR
LABS991PRT16 LABS991PRT15 LABS991PRT14 LABS991PRT13 LABS991001024 LABS991PRT12 LABS991PRT11 LABS991001096 LABS991STR11 LABS991STR12
MSIA pipette tips
Improve hybrid MS bio-analysis with Thermo Scientific™ MSIA™ D.A.R.T.'S™ for the Versette and Novus i Platform, an exclusive Thermo Fisher Scientific workflow solution for proteins and peptides.
  • Enables enrichment and concentratation of protein of interest down to femtomole levels
  • Streamlines mass spectrometry targeted proteomic quantification workflow
  • High specificity: Immunoaffinity column captures protein of interest with improved signal-to-noise ratio
  • Better sensitivity: Superior protein recovery compared to conventional bead-based method
Enabled through proprietary MSIA D.A.R.T.'S™ technologies, these workflows have demonstrated pg/ml level detection while providing simultaneous qualitative/quantitative data content from complex biological matrix, such as human plasma. Ideal for biologic and biomarker bio-analysis, MSIA provides innovative enablement in the areas of PKPD, DMPK, ADME, Biotransformation (DAR and payload determination), as well as routine biomarker analysis.
For use with Thermo Scientific™ Versette™ Liquid Handling Platform (fully automated method) or Thermo Scientific™ Finnpipette™ Novus i™ Multichannel pipettors (semi-automated method).