Protein MW markers, BlueStep™ high range, pre-stained
Lieferant: VWR Life Science
K974-0.5MLEA 0 EUR
VWRCK974-0.5ML
Protein MW markers, BlueStep™ high range, pre-stained
BlueStep™ protein molecular weight marker, high range, pre-stained has 5 bands ranging from 34,0 to 200,0 kDa. Pre-stained bands are easily transferred to blotting membranes for use in Western blotting. There is sufficient material for 50 assays.
  • 5 bands ranging from 34,0 to 200,0 kDa
  • Monitor real-time running conditions during electrophoresis