Precast gels, PhastGel® Dry IEF
Lieferant: Cytiva
17-0677-01EA 229 EUR
GEHE17-0677-01
Precast gels, PhastGel® Dry IEF
PhastGel® Dry IEF combines the freedom and versatility of a homemade gel with the convenience, reproducibility, increased safety and speed of a precast gel.
  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing, native, and isoelectric focusing gels
  • Save time and improve reproducibility with precast gels, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready to use silver staining kits
The gel is easily rehydrated in a rehydration cassette.