illustra™ MicroSpin™ S-200 HR columns
Lieferant: Cytiva
illustra™
27-5120-01EA 253 EUR
27-5120-01
illustra™ MicroSpin™ S-200 HR columns
Nukleinsäure-Reagenzien Nucleic Acid Purification Kits and Reagents
illustra™ MicroSpin™ S-200 HR columns allow rapid purification of labelled single-stranded or double-stranded DNA fragments >50 bases in length in a volume of 51 to 100 μl.