Anion exchange chromatography column, Mono Q HR
Lieferant: Cytiva
17-0506-01EA 19720 EUR
17-0506-01
Anion exchange chromatography column, Mono Q HR
Chromatographiesäulen
Mono Q HR 16/10 is a strong anion exchanger prepacked with MonoBeads.
  • Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability.
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers.
  • Compatible with ÄKTA design, FPLC System, and other high-performance LC systems.
The column is an excellent choice for the polishing step in purification of proteins, peptides, and other biomolecules when high purity is required. The 20 mL bed volume of the column offers high capacity.