Anion exchange chromatography columns, HiLoad™ Q Sepharose™ HP
Lieferant: Cytiva
17-1066-01EA 1935 EUR
GEHE17-1066-01
Anion exchange chromatography columns, HiLoad™ Q Sepharose™ HP
HiLoad™ 16/10 and 26/10 Q Sepharose™ High Performance is a prepacked, strong anion exchanger XK column designed for high resolution preparative ion exchange chromatography separations.
  • For high-performance preparative scale separations where high efficiency and high resolution are desired
  • Q is a strong ion exchanger
  • Highly cross-linked agarose provides excellent chemical and physical stability
  • Higher resolution than HiPrep Sepharose Fast Flow ion exchangers