Fusion Solo S chemiluminescence imaging system offers high resolution imaging and reliable quantitation, ideal for Western blotting, bioluminescence and luciferase assay applications.
- Monochrome scientific grade 16-bit CCD camera, 4,2/10,0 megapixel (Fusion Solo 7S) or 6,0/18,0 megapixel (Fusion Solo 6S)
- F0,84 manual lens designed for chemiluminescence
- Camera cooling to lower background noise
- Up to 8×8 binning to detect weak signals
- Intuitive capture software with flat field correction, auto exposure/manual exposure/video mode
Lieferumfang: Transilluminator and filters must be ordered separately. For more information about options and accessories available with this system please contact your local VWR sales office.