Chemiluminescence imaging system, Fusion Solo S
Lieferant: Vilber Lourmat
VILB121176001EA 24550 EUR
VILB121176001 VILB121172001
Chemiluminescence imaging system, Fusion Solo S
Fusion Solo S chemiluminescence imaging system offers high resolution imaging and reliable quantitation, ideal for Western blotting, bioluminescence and luciferase assay applications.
  • Monochrome scientific grade 16-bit CCD camera, 4,2/10,0 megapixel (Fusion Solo 7S) or 6,0/18,0 megapixel (Fusion Solo 6S)
  • F0,84 manual lens designed for chemiluminescence
  • Camera cooling to lower background noise
  • Up to 8×8 binning to detect weak signals
  • Intuitive capture software with flat field correction, auto exposure/manual exposure/video mode
Lieferumfang: Transilluminator and filters must be ordered separately. For more information about options and accessories available with this system please contact your local VWR sales office.