Liquid scintillation cocktails
Lieferant: Cytiva
NOCS104EA 508 EUR
GEHENOCS104
Liquid scintillation cocktails
BCS for aqueous samples: Biodegradable scintillant. Efficiencies with ³H of 30 - 40%. Low odour, low toxicity and a high flashpoint (>150 °C) makes it less hazardous to the user and the environment.
OCS for non aqueous samples: High efficiency xylene-based scintillant. Efficiency with ³H of >50%. Flashpoint 26,7 °C.
OCS for non aqueous samples: High efficiency xylene-based scintillant. Efficiency with ³H of >50%. Flashpoint 26,7 °C.