Streptavidin HP SpinTrap™ buffer kit
Lieferant: Cytiva
28-9135-68EA 201 EUR
GEHE28-9135-68
Streptavidin HP SpinTrap™ buffer kit
Streptavidin HP SpinTrap™ Buffer kit comprises pre-made buffers optimised for the affinity-based enrichment of a target protein/immunoprecipitation using Streptavidin HP SpinTrap columns.
  • High quality buffers
  • All buffer components listed and defined
  • Convenient format - stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment
  • Sufficient for performing 16 reactions
The buffers provided in the kit are optimised for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with Streptavidin HP SpinTrap™ columns.
Lieferumfang: Kit includes 10X Binding/Washing Buffer [TBS]: 0,5 M Tris, 1,5 M NaCl, pH 7.5; 10X Elution Buffer: 1 M glycine-HCl, pH 2.9; Blocking Buffer: 2 mM biotin; 50 mM Tris, 0,15 M NaCl, pH 7.5.