Handheld thermo-hygrometer, HygroPalm 21
Lieferant: Rotronic
620-2750EA 398 EUR
ROTRHP21
Handheld thermo-hygrometer, HygroPalm 21
The HP21 with fixed M1-R / PT 100 class A probe (L×Ø: 80×15 mm) and PE filter is designed for humidity and temperature measurements. It is suitable for applications such as mobile inspection and random tests in HVAC, the pharmaceutical, food and paper industries, meteorology and building management system. It provides an important indicator for the shelf life of products and it strongly influences the occurrence and growth of microorganisms.
  • USB interface to the PC
  • Dew/frost points calculation
  • Highly repeatable rapid response
  • Auto-diagnostics with error correction and trend indicators
  • User calibration adjustment via keypad with the appropriate standards
Conforms to FDA 21 CFR Part 11 and GAMP compliant.
Zertifizierungen: Conforms to FDA 21 CFR Part 11 and GAMP compliant.
Lieferumfang: Supplied with a nine-volt battery, a low battery indicator and a factory adjustement certificate.