Accessories
Lieferant: PerkinElmer
PERCN9304555EA 1080 EUR
PERCN9304555 PERCN9304552 PERCN9304553 PERCN9304557
Accessories
These AxION® separation column probe holders facilitate chromatography directly at the ionisation source.
  • Minimises dead volume effects in the system for superior signal-to-noise ratios
  • Enhanced sensitivity and narrower peaks