Thistle Trichter, Azlon®
Lieferant: SciLabware
221-0046EA 190 EUR
AZLOFWC220
Thistle Trichter, Azlon®
Thistle shaped funnel, made of PP, with detachable long stem.
  • Stem can be cut to size
  • Good chemical resistance
  • Good temperature resistance