Ruthenium, schwarz, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL326712-1GEA 102 EUR
SIAL326712-1G
Ruthenium, schwarz, Sigma-Aldrich®
Ruthenium
Formel: Ru
Molekulargewicht: 101,07 g/mol
MDL Number: MFCD00011207
CAS-Nummer: 7440-18-8
EINECS: 231-127-1
UN-Nummer: 3089
ADR: 4.1,III
Merck Index: 12,08452

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR