3-(1H-Pyrazol-1-yl)benzaldehyd

Lieferant: Thermo Scientific

MAYBCC32904DAEA 0 EUR
MAYBCC32904DA
3-(1H-Pyrazol-1-yl)benzaldehyd
3-(1H-Pyrazol-1-yl)benzaldehyd
Formel: C₁₀H₈N₂O
Molekulargewicht: 172,18 g/mol
MDL Number: MFCD06740321
CAS-Nummer: 852227-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR