3'-Chlor-4'-ethoxy-5'-fluoracetophenon 97%, JRD
|
Achtung
|
H32546.03EA
22.8
EUR
H32546.03
H32546.06
3'-Chlor-4'-ethoxy-5'-fluoracetophenon 97%, JRD
3'-Chlor-4'-ethoxy-5'-fluoracetophenon
Formel:
C₁₀H₁₀ClFO₂ Molekulargewicht: 216,64 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832347 CAS-Nummer: 1017779-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...