D(-)-Diisopropyltartrat

Lieferant: Molekula

MOLEM67988409EA 0 EUR
MOLEM67988409
D(-)-Diisopropyltartrat
D(-)-Diisopropyltartrat
Formel: C₁₀H₁₈O₆
Molekulargewicht: 234,25 g/mol
MDL Number: MFCD00008876
CAS-Nummer: 62961-64-2
EINECS: 263-771-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR