5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL433853-5GEA
255
EUR
SIAL433853-5G
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd
Formel:
C₉H₁₁ClN₂O Molekulargewicht: 220,65 g/mol |
MDL Number:
MFCD00100755 CAS-Nummer: 947-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...