Diclofenac-Natrium
Lieferant: EHRENSTORFER GMBH
EHERC12537500EA
78.2
EUR
EHERC12537500
Diclofenac-Natrium
Diclofenac-Natrium
Formel:
C₁₄H₁₀Cl₂NNaO₂ Molekulargewicht: 318,1 g/mol |
MDL Number:
MFCD00082251 CAS-Nummer: 15307-79-6 EINECS: 239-346-4 UN-Nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...