Zinn(II)phthalocyanin
Lieferant: Thermo Scientific
|
Gefahr
|
39607.02EA
81.9
EUR
39607.02
Zinn(II)phthalocyanin
Tin(II) phthalocyanine
Formel:
C₃₂H₁₆N₈Sn Molekulargewicht: 631,233 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058867 CAS-Nummer: 15304-57-1 UN-Nummer: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...