Carbamazepin
Lieferant: FITZGERALD INDUSTRIES INT.
FITZ20-CS06EA
0
EUR
FITZ20-CS06
Carbamazepin
Carbamazepin
Formel:
C₁₅H₁₂N₂O Molekulargewicht: 236,27 g/mol |
MDL Number:
MFCD00005073 CAS-Nummer: 298-46-4 EINECS: 206-062-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...