D-Glucoronic acid

Lieferant: Molekula

MOLEM24989192EA 0 EUR
MOLEM24989192
D-Glucoronic acid
D-Glucoronic acid
Formel: C₆H₁₀O₇
MDL Number: MFCD00166981
CAS-Nummer: 6556-12-3
EINECS: 229-486-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR