Diosmin

Lieferant: Molekula

MOLEM14577662EA 0 EUR
MOLEM14577662
Diosmin
Diosmin
Formel: C₂₈H₃₂O₁₅
Molekulargewicht: 608,5 g/mol
MDL Number: MFCD00009772
CAS-Nummer: 520-27-4
EINECS: 208-289-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR